N-(3,6-Dimethylpyridin-2-yl)pivalamide ≥98%

Lieferant: Apollo Scientific

OR370050-1G
APOSOR370050-1GEA 993 EUR
APOSOR370050-1G
N-(3,6-Dimethylpyridin-2-yl)pivalamide ≥98%
N-(3,6-Dimethylpyridin-2-yl)pivalamide
Formel: C₁₂H₁₈N₂O
Molecular Weight: 206,284 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD19443197
CAS-Nummer: 1313762-34-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR