4-tert-Butyl-2-methoxybenzaldehyde ≥98%

Lieferant: Apollo Scientific

OR370051-1G OR370051-5G
APOSOR370051-1GEA 447 EUR
APOSOR370051-1G APOSOR370051-5G
4-tert-Butyl-2-methoxybenzaldehyde ≥98%
4-tert-Butyl-2-methoxybenzaldehyde
Formel: C₁₂H₁₆O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD18642730
CAS-Nummer: 53535-88-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR