1-((4R,5R)-2,2-dimethyl-5-vinyl-1,3-dioxolan-4-yl)ethanone ≥98%

Lieferant: Apollo Scientific

OR370052-1G
APOSOR370052-1GEA 789 EUR
APOSOR370052-1G
1-((4R,5R)-2,2-dimethyl-5-vinyl-1,3-dioxolan-4-yl)ethanone ≥98%
1-((4R,5R)-2,2-dimethyl-5-vinyl-1,3-dioxolan-4-yl)ethanone
Formel: C₉H₁₄O₃
Molecular Weight: 170,206 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD19443200
CAS-Nummer: 1042237-51-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR