3-(Phenylimino)pentane-2,4-dione ≥98%
Lieferant: Apollo Scientific
OR370053-1G
APOSOR370053-1GEA
300
EUR
APOSOR370053-1G
3-(Phenylimino)pentane-2,4-dione ≥98%
3-(Phenylimino)pentane-2,4-dione
Formel:
C₁₁H₁₁NO₂ Molecular Weight: 189,211 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD18642735 CAS-Nummer: 83325-65-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...