5-Acetyl-5-azaspiro[2.4]heptan-7-one ≥98%
Lieferant: Apollo Scientific
OR370054-100MG
APOSOR370054-100MGEA
910
EUR
APOSOR370054-100MG
5-Acetyl-5-azaspiro[2.4]heptan-7-one ≥98%
5-Acetyl-5-azaspiro[2.4]heptan-7-one
Formel:
C₈H₁₁NO₂ Molecular Weight: 153,178 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD18642737 CAS-Nummer: 400841-12-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...