N-(8-Methyl-8-azabicyclo[3.2.1]octan-3-yl)benzamide ≥98%

Lieferant: Apollo Scientific

OR370055-100MG
APOSOR370055-100MGEA 525 EUR
APOSOR370055-100MG
N-(8-Methyl-8-azabicyclo[3.2.1]octan-3-yl)benzamide ≥98%
N-(8-Methyl-8-azabicyclo[3.2.1]octan-3-yl)benzamide
Formel: C₁₅H₂₀N₂O
Molecular Weight: 244,332 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00471181
CAS-Nummer: 329738-53-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR