3-Ethyl-2,3-dimethyl-1,2-oxaziridine ≥98%

Lieferant: Apollo Scientific

OR370058-25MG
APOSOR370058-25MGEA 381 EUR
APOSOR370058-25MG
3-Ethyl-2,3-dimethyl-1,2-oxaziridine ≥98%
3-Ethyl-2,3-dimethyl-1,2-oxaziridine
Formel: C₅H₁₁NO
Molecular Weight: 101.147 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18642810
CAS-Nummer: 3400-16-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR