2-(3-Amino-5-methoxyphenyl)ethanol ≥98%

Lieferant: Apollo Scientific

OR370059-100MG OR370059-1G
APOSOR370059-100MGEA 362 EUR
APOSOR370059-100MG APOSOR370059-1G
2-(3-Amino-5-methoxyphenyl)ethanol ≥98%
2-(3-Amino-5-methoxyphenyl)ethanol
Formel: C₉H₁₃NO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD18642813
CAS-Nummer: 354512-41-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR