2-Oxo-2,3,4,5-tetrahydro-1H-benzo[b]azepine-8-carbonitrile ≥98%

Lieferant: Apollo Scientific

OR370060-100MG
APOSOR370060-100MGEA 1210 EUR
APOSOR370060-100MG
2-Oxo-2,3,4,5-tetrahydro-1H-benzo[b]azepine-8-carbonitrile ≥98%
2-Oxo-2,3,4,5-tetrahydro-1H-benzo[b]azepine-8-carbonitrile
Formel: C₁₁H₁₀N₂O
Molecular Weight: 186,21 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18642821
CAS-Nummer: 903557-04-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR