2-(2,3-Dihydrobenzofuran-5-yl)-1-morpholinoethanethione ≥98%

Lieferant: Apollo Scientific
Achtung

OR370062-25MG
APOSOR370062-25MGEA 353 EUR
APOSOR370062-25MG
2-(2,3-Dihydrobenzofuran-5-yl)-1-morpholinoethanethione ≥98%
2-(2,3-Dihydrobenzofuran-5-yl)-1-morpholinoethanethione
Formel: C₁₄H₁₇NO₂S
Molecular Weight: 263,355 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18642881
CAS-Nummer: 97483-11-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR