7-Chloro-2-(chloromethyl)-6-methylquinazolin-4(3H)-one ≥98%

Lieferant: Apollo Scientific

OR370066-100MG
APOSOR370066-100MGEA 688 EUR
APOSOR370066-100MG
7-Chloro-2-(chloromethyl)-6-methylquinazolin-4(3H)-one ≥98%
7-Chloro-2-(chloromethyl)-6-methylquinazolin-4(3H)-one
Formel: C₁₀H₈Cl₂N₂O
Molecular Weight: 243,089 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12828614
CAS-Nummer: 289686-83-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR