2-Cyanoquinuclidine ≥98%

Lieferant: Apollo Scientific

OR370068-100MG
APOSOR370068-100MGEA 626 EUR
APOSOR370068-100MG
2-Cyanoquinuclidine ≥98%
2-Cyanoquinuclidine
Formel: C₈H₁₂N₂
Lagertemperatur: Raumtemperatur
MDL: MFCD22571233
CAS-Nummer: 90196-91-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR