2-(2-Methoxyphenyl)-1H-imidazole ≥98%

Lieferant: Apollo Scientific

OR370070-1G
APOSOR370070-1GEA 688 EUR
APOSOR370070-1G
2-(2-Methoxyphenyl)-1H-imidazole ≥98%
2-(2-Methoxyphenyl)-1H-imidazole
Formel: C₁₀H₁₀N₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD08668846
CAS-Nummer: 52091-35-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR