1-(3,5-Dichloropyridin-4-yl)ethanol ≥98%

Lieferant: Apollo Scientific

OR370074-1G
APOSOR370074-1GEA 104 EUR
APOSOR370074-1G
1-(3,5-Dichloropyridin-4-yl)ethanol ≥98%
1-(3,5-Dichloropyridin-4-yl)ethanol
Formel: C₇H₇Cl₂NO
Molecular Weight: 192.043 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD23163275
CAS-Nummer: 1254473-66-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR