(E)-2-(1,2,6,7-Tetrahydro-8H-indeno[5,4-b]furan-8-ylidene)acetonitrile ≥98%

Lieferant: Apollo Scientific

OR370075-1G
APOSOR370075-1GEA 95.7 EUR
APOSOR370075-1G
(E)-2-(1,2,6,7-Tetrahydro-8H-indeno[5,4-b]furan-8-ylidene)acetonitrile ≥98%
(E)-2-(1,2,6,7-Tetrahydro-8H-indeno[5,4-b]furan-8-ylidene)acetonitrile
Formel: C₁₃H₁₁NO
Molecular Weight: 197,233 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09955086
CAS-Nummer: 196597-79-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR