3-Methyl-4-(piperidin-4-yl)aniline ≥98%

Lieferant: Apollo Scientific

OR370079-1G
APOSOR370079-1GEA 1660 EUR
APOSOR370079-1G
3-Methyl-4-(piperidin-4-yl)aniline ≥98%
3-Methyl-4-(piperidin-4-yl)aniline
Formel: C₁₂H₁₈N₂
Molecular Weight: 190,285 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD17486190
CAS-Nummer: 1260864-79-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR