3-Bromo-2-methyl-4-quinolinol ≥95%

Lieferant: Apollo Scientific

OR370082-1G
APOSOR370082-1GEA 340 EUR
APOSOR370082-1G
3-Bromo-2-methyl-4-quinolinol ≥95%
3-Bromo-2-methyl-4-quinolinol
Formel: C₁₀H₈BrNO
Molecular Weight: 238,081 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD13689063
CAS-Nummer: 65673-88-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR