3-Iodo-2-methylquinolin-4-ol ≥98%

Lieferant: Apollo Scientific

OR370083-1G
APOSOR370083-1GEA 1640 EUR
APOSOR370083-1G
3-Iodo-2-methylquinolin-4-ol ≥98%
3-Iodo-2-methylquinolin-4-ol
Formel: C₁₀H₈INO
Molecular Weight: 285,081 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD24368411
CAS-Nummer: 64965-49-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR