1-((Trimethylsilyl)ethynyl)cyclopropane-1-carboxylic acid ≥98%

Lieferant: Apollo Scientific

OR370086-1G
APOSOR370086-1GEA 302 EUR
APOSOR370086-1G
1-((Trimethylsilyl)ethynyl)cyclopropane-1-carboxylic acid ≥98%
1-((Trimethylsilyl)ethynyl)cyclopropane-1-carboxylic acid
Formel: C₉H₁₄O₂Si
Molecular Weight: 182,292 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD20660917
CAS-Nummer: 1268810-07-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR