3-(7-Bromo-2,3-dihydrobenzofuran-5-yl)propanoic acid ≥98%
Lieferant: Apollo Scientific
OR370088-1G
APOSOR370088-1GEA
910
EUR
APOSOR370088-1G
3-(7-Bromo-2,3-dihydrobenzofuran-5-yl)propanoic acid ≥98%
3-(7-Bromo-2,3-dihydrobenzofuran-5-yl)propanoic acid
Formel:
C₁₁H₁₁BrO₃ Molecular Weight: 271,107 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD29053188 CAS-Nummer: 196597-68-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...