2,3,6,7-Tetrahydro-1H-azepine hydrochloride ≥98%

Lieferant: Apollo Scientific

OR370090-250MG
APOSOR370090-250MGEA 724 EUR
APOSOR370090-250MG
2,3,6,7-Tetrahydro-1H-azepine hydrochloride ≥98%
2,3,6,7-Tetrahydro-1H-azepine hydrochloride
Formel: C₆H₁₁N·ClH
Molecular Weight: 133,619 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD17676921
CAS-Nummer: 1263282-12-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR