3-Chloro-N-(4-piperidinylmethyl)benzamide hydrochloride ≥98%

Lieferant: Apollo Scientific

OR370093-1G
APOSOR370093-1GEA 0 EUR
APOSOR370093-1G
3-Chloro-N-(4-piperidinylmethyl)benzamide hydrochloride ≥98%
3-Chloro-N-(4-piperidinylmethyl)benzamide hydrochloride
Formel: C₁₃H₁₇ClN₂O·ClH
Lagertemperatur: Raumtemperatur
MDL: MFCD30536406
CAS-Nummer: 1584066-83-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR