4-Bromo-2,3-Dihydroxybenzaldehyde ≥98%

Lieferant: Apollo Scientific

OR370094-500MG
APOSOR370094-500MGEA 1030 EUR
APOSOR370094-500MG
4-Bromo-2,3-Dihydroxybenzaldehyde ≥98%
4-Bromo-2,3-Dihydroxybenzaldehyde
Formel: C₇H₅BrO₃
Molecular Weight: 217,017 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD30738024
CAS-Nummer: 73275-98-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR