N-[1-(2-Naphthalenyl)ethyl]formamide 98

Lieferant: Apollo Scientific

OR370095-1G
APOSOR370095-1GEA 545 EUR
APOSOR370095-1G
N-[1-(2-Naphthalenyl)ethyl]formamide 98
N-[1-(2-Naphthalenyl)ethyl]formamide
Formel: C₁₃H₁₃NO
Molecular Weight: 199.248 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD13250145
CAS-Nummer: 273384-77-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR