N-[4-Chloro-2-methoxy-5-(1-methylethenyl)phenyl]acetamide ≥98%

Lieferant: Apollo Scientific

OR370096-1G
APOSOR370096-1GEA 1810 EUR
APOSOR370096-1G
N-[4-Chloro-2-methoxy-5-(1-methylethenyl)phenyl]acetamide ≥98%
N-[4-Chloro-2-methoxy-5-(1-methylethenyl)phenyl]acetamide
Formel: C₁₂H₁₄ClNO₂
Molecular Weight: 239,698 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD30730232
CAS-Nummer: 1629269-89-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR