Benzo[d]oxazol-5-ol ≥98%

Lieferant: Apollo Scientific

OR370097-1G OR370097-250MG
APOSOR370097-1GEA 0 EUR
APOSOR370097-1G APOSOR370097-250MG
Benzo[d]oxazol-5-ol ≥98%
Benzo[d]oxazol-5-ol
Formel: C₇H₅NO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD11110248
CAS-Nummer: 180716-28-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR