1-(1-Methylcyclopropyl)ethanamine hydrochloride ≥98%

Lieferant: Apollo Scientific

OR370099-100MG
APOSOR370099-100MGEA 847 EUR
APOSOR370099-100MG
1-(1-Methylcyclopropyl)ethanamine hydrochloride ≥98%
1-(1-Methylcyclopropyl)ethanamine hydrochloride
Formel: C₆H₁₃N·ClH
Molecular Weight: 135,635 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09971518
CAS-Nummer: 54343-93-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR