3-(3-Bromophenyl)propanal 95%

Lieferant: Apollo Scientific

OR370108-1G
APOSOR370108-1GEA 509 EUR
APOSOR370108-1G
3-(3-Bromophenyl)propanal 95%
3-(3-Bromophenyl)propanal
Formel: C₉H₉BrO
Molecular Weight: 213,071 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09028602
CAS-Nummer: 210115-30-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR