(R)-1-Azabicyclo[2.2.2]octane-2-carboxylic acid, hydrochloride 95%
Lieferant: Apollo Scientific
OR370109-250MG
APOSOR370109-250MGEA
1210
EUR
APOSOR370109-250MG
(R)-1-Azabicyclo[2.2.2]octane-2-carboxylic acid, hydrochloride 95%
(R)-1-Azabicyclo[2.2.2]octane-2-carboxylic acid, hydrochloride
Formel:
C₈H₁₃NO₂·ClH Molecular Weight: 191,655 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD30207805 CAS-Nummer: 94160-98-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...