4-Bromo-N-methyl-1H-pyrrolo[2,3-b]pyridin-6-amine 95%

Lieferant: Apollo Scientific

OR370118-250MG
APOSOR370118-250MGEA 243 EUR
APOSOR370118-250MG
4-Bromo-N-methyl-1H-pyrrolo[2,3-b]pyridin-6-amine 95%
4-Bromo-N-methyl-1H-pyrrolo[2,3-b]pyridin-6-amine
Formel: C₈H₈BrN₃
Molecular Weight: 226,073 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28346930
CAS-Nummer: 1935288-47-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR