5-Bromo-3-(2-methyl-4-thiazolyl)-1H-Pyrrolo[2,3-b]pyridine 95%
Lieferant: Apollo Scientific
OR370119-100MG
APOSOR370119-100MGEA
376
EUR
APOSOR370119-100MG
5-Bromo-3-(2-methyl-4-thiazolyl)-1H-Pyrrolo[2,3-b]pyridine 95%
5-Bromo-3-(2-methyl-4-thiazolyl)-1H-Pyrrolo[2,3-b]pyridine
Formel:
C₁₁H₈BrN₃S Molecular Weight: 294,17 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD15529159 CAS-Nummer: 1046793-78-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...