6-Bromo-8-methoxyisoquinoline ≥97%

Lieferant: Apollo Scientific

OR370121-100MG
APOSOR370121-100MGEA 368 EUR
APOSOR370121-100MG
6-Bromo-8-methoxyisoquinoline ≥97%
6-Bromo-8-methoxyisoquinoline
Formel: C₁₀H₈BrNO
Molecular Weight: 238,081 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22124579
CAS-Nummer: 1415564-97-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR