6-Hydroxy-1H-indole-3-carbaldehyde 98%

Lieferant: Apollo Scientific

OR370135-250MG
APOSOR370135-250MGEA 777 EUR
APOSOR370135-250MG
6-Hydroxy-1H-indole-3-carbaldehyde 98%
6-Hydroxy-1H-indole-3-carbaldehyde
Formel: C₉H₇NO₂
Molecular Weight: 161,157 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11110391
CAS-Nummer: 192184-71-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR