5-Bromofuran-2-carbonitrile 95%

Lieferant: Apollo Scientific

OR370139-1G
APOSOR370139-1GEA 46.4 EUR
APOSOR370139-1G
5-Bromofuran-2-carbonitrile 95%
5-Bromofuran-2-carbonitrile
Formel: C₅H₂BrNO
Lagertemperatur: Raumtemperatur
MDL: MFCD03410206
CAS-Nummer: 4915-06-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR