5-Brom-2-chlorisonicotinsäure 97%
Lieferant: Apollo Scientific
|
Achtung
|
OR3702-100G
OR3702-25G
APOSOR3702-100GEA
221
EUR
APOSOR3702-100G
APOSOR3702-25G
5-Brom-2-chlorisonicotinsäure 97%
5-Brom-2-chlorisonicotinsäure
Formel:
C₆H₃BrClNO₂ Schmelzpunkt: 220…224 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD07375114 CAS-Nummer: 886365-31-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...