5-Brom-2-chlor-4-methoxypyrimidin

Lieferant: Apollo Scientific
Achtung

OR3706-1G
APOSOR3706-1GEA 21.9 EUR
APOSOR3706-1G
5-Brom-2-chlor-4-methoxypyrimidin
5-Bromo-2-chloro-4-methoxypyrimidine
Formel: C₅H₄BrClN₂O
Molecular Weight: 223,4600 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD03001373
CAS-Nummer: 57054-92-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR