1-(2-Brombenzyl)piperidin

Lieferant: Apollo Scientific
Achtung

OR3711-1G
APOSOR3711-1GEA 92.8 EUR
APOSOR3711-1G
1-(2-Brombenzyl)piperidin
1-(2-Brombenzyl)piperidin
Formel: C₁₂H₁₆BrN
Lagertemperatur: Kühlschrank
CAS-Nummer: 1459-22-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR