3-Amino-4-hydroxypyridine

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Amino-4-pyridinol

OR3719-1G
APOSOR3719-1GEA 21.9 EUR
APOSOR3719-1G
3-Amino-4-hydroxypyridine
3-Amino-4-hydroxypyridine
Formel: C₅H₆N₂O
Molecular Weight: 110,12 g/mol
Schmelzpunkt: 145…149 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD04114230
CAS-Nummer: 6320-39-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR