3-Amino-2-brom-5-chlorpyridin 97%
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
2-Brom-5-chlor-3-pyridylamin
OR3720-1G
APOSOR3720-1GEA
21.9
EUR
APOSOR3720-1G
3-Amino-2-brom-5-chlorpyridin 97%
3-Amino-2-bromo-5-chloropyridine
Formel:
C₅H₄BrClN₂ Molecular Weight: 207,46 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD07368976 CAS-Nummer: 90902-83-3 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...