C-(1-Benzylpyrrolidin-3-yl)methylamine

Lieferant: Apollo Scientific
Achtung

Synonyme: (1-Benzylpyrrolidin-3-yl)methanamine

OR3729-1G
APOSOR3729-1GEA 0 EUR
APOSOR3729-1G
C-(1-Benzylpyrrolidin-3-yl)methylamine
C-(1-Benzylpyrrolidin-3-yl)methylamine
Formel: C₁₂H₁₈N₂
Molecular Weight: 190,29 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06411056
CAS-Nummer: 93138-61-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR