2-Phenylnicotinic acid
Lieferant: Apollo Scientific
|
Achtung
|
OR3732-250MG
APOSOR3732-250MGEA
27.7
EUR
APOSOR3732-250MG
2-Phenylnicotinic acid
2-Phenylnicotinic acid
Formel:
C₁₂H₉NO₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD03069417 CAS-Nummer: 33421-39-5 EINECS: 251-510-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...