4-(Methylsulfonyl)phenylboronsäure

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-(Methylsulphonyl)benzeneboronic acid, 4-(Methanesulphonyl)phenylboronic acid

OR3734-1G
APOSOR3734-1GEA 21.9 EUR
APOSOR3734-1G
4-(Methylsulfonyl)phenylboronsäure
4-(Methylsulfonyl)phenylboronsäure
Formel: C₇H₉BO₄S
Molecular Weight: 200,02 g/mol
Schmelzpunkt: 289…293 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01630820
CAS-Nummer: 149104-88-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR