5-Nitro-2-thiophencarbonsäure
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
5-Nitrothiophene-2-carboxylic acid
OR3736-1G
APOSOR3736-1GEA
29
EUR
APOSOR3736-1G
5-Nitro-2-thiophencarbonsäure
5-Nitro-2-thiophenecarboxylic acid
Formel:
C₅H₃NO₄S Molecular Weight: 173,14 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00159552 CAS-Nummer: 6317-37-9 EINECS: 228-654-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...