[1-(Sulfanylmethyl)cyclopropyl]essigsäure 97%
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
[1-(Thiomethyl)cycloprop-1-yl]acetic acid, 2-(1-(Mercaptomethyl)cyclopropyl)acetic acid
OR3739-1G
APOSOR3739-1GEA
21.9
EUR
APOSOR3739-1G
[1-(Sulfanylmethyl)cyclopropyl]essigsäure 97%
1-(Mercaptomethyl)cyclopropaneacetic acid
Formel:
C₆H₁₀O₂S Molecular Weight: 146,20 g/mol Schmelzpunkt: 42…46 °C Flammpunkt: >110 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03411600 CAS-Nummer: 162515-68-6 EINECS: 420-240-3 UN: 3261 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...