N,N-Bis(isopropyl)benzylamine

Lieferant: Apollo Scientific
Achtung

OR3756-25G
APOSOR3756-25GEA 0 EUR
APOSOR3756-25G
N,N-Bis(isopropyl)benzylamine
N,N-Bis(isopropyl)benzylamine
Formel: C₁₃H₂₁N
Dichte: 0,9 g/cm³ (25 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD07368226
CAS-Nummer: 34636-09-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR