2-Chlor-4-methyl-5-pyridinboronsäure 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: (6-Chloro-4-methylpyridin-3-yl)boronic acid, 2-Chloro-4-methylpyridine-5-boronic acid

OR3766-1G
APOSOR3766-1GEA 21.9 EUR
APOSOR3766-1G
2-Chlor-4-methyl-5-pyridinboronsäure 98%
2-Chlor-4-methyl-5-pyridinboronsäure
Formel: C₆H₇BClNO₂
Molecular Weight: 171,3900 g/mol
Schmelzpunkt: 105…107 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD07368882
CAS-Nummer: 913836-08-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR