4-(Benzyl(ethyl)carbamoyl)phenylboronsäure 98%

Lieferant: Apollo Scientific
Achtung

OR3772-1G
APOSOR3772-1GEA 69.6 EUR
APOSOR3772-1G
4-(Benzyl(ethyl)carbamoyl)phenylboronsäure 98%
4-(Benzyl(ethyl)carbamoyl)phenylboronsäure
Formel: C₁₆H₁₈BNO₃
Schmelzpunkt: 140…145 °C
Lagertemperatur: Kühlschrank
MDL: MFCD09027215
CAS-Nummer: 913835-41-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR