4-(3-Methoxypropylcarbamoyl)phenylboronsäure 98%

Lieferant: Apollo Scientific
Achtung

OR3774-1G
APOSOR3774-1GEA 60.9 EUR
APOSOR3774-1G
4-(3-Methoxypropylcarbamoyl)phenylboronsäure 98%
4-(3-Methoxypropylcarbamoyl)phenylboronsäure
Formel: C₁₁H₁₆BNO₄
Lagertemperatur: Kühlschrank
MDL: MFCD09027225
CAS-Nummer: 913835-85-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR