3,4-Dimethylphenylhydraziniumchlorid 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 3,4-Dimethylphenylhydrazinhydrochlorid, 3,4-Dimethylphenylhydrazin Hydrochlorid, 1-(3,4-Dimethylphenyl)hydrazin Hydrochlorid

OR3775-100G
APOSOR3775-100GEA 62.5 EUR
APOSOR3775-100G
3,4-Dimethylphenylhydraziniumchlorid 98%
3,4-Dimethylphenylhydraziniumchlorid
Formel: C₈H₁₃ClN₂
Molecular Weight: 172.66 g/mol
Schmelzpunkt: 196 °C (Zers.)
Lagertemperatur: Kühlschrank
MDL: MFCD00052270
CAS-Nummer: 60481-51-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR