2,7-Dimethylnaphthalin
Lieferant: Apollo Scientific
OR3785-1G
APOSOR3785-1GEA
84.1
EUR
APOSOR3785-1G
2,7-Dimethylnaphthalin
2,7-Dimethylnaphthalene
Formel:
C₁₂H₁₂ Molecular Weight: 156,23 g/mol Siedepunkt: 262…264 °C Schmelzpunkt: 96…98 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00004121 CAS-Nummer: 582-16-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...